Gene Transcripts
- Gene Symbol
- CTRB2
- Entrez Gene ID
- 440387
- Gene Name
- chymotrypsinogen B2
- Gene Aliases
- Chromosome Location
- Chr. 16: 75204096 - 75207174 on Build GRCh38
- UniGene ID
- Hs.610926
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124403.1 |
- |
4 |
2197 |
|
- Gene Symbol
- CTRB1
- Entrez Gene ID
- 1504
- Gene Name
- chymotrypsinogen B1
- Gene Aliases
- CTRB
- Chromosome Location
- Chr. 16: 75218986 - 75224924 on Build GRCh38
- UniGene ID
- Hs.610926
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC005385.1 |
- |
7 |
807 |
|
BT007356.1 |
- |
6 |
765 |
|
M24400.1 |
- |
6 |
781 |
|
View Full Product Details