Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290223.1 | NP_001277152.1 | 21 | 2231 | |
NM_001380.4 | NP_001371.1 | 21 | 2168 | ||
XM_011539422.2 | XP_011537724.1 | 21 | 2209 | ||
XM_011539424.1 | XP_011537726.1 | 21 | 2119 | ||
XM_017015813.1 | XP_016871302.1 | 21 | 2141 | ||
XM_017015814.1 | XP_016871303.1 | 21 | 2203 | ||
XM_017015815.1 | XP_016871304.1 | 21 | 2226 | ||
XM_017015816.1 | XP_016871305.1 | 5 | 498 | ||
XM_017015817.1 | XP_016871306.1 | 21 | 2203 | ||
GenBank mRNA | BC146857.1 | - | 21 | 2121 | |
CX163251.1 | - | 5 | 472 | ||
D50857.1 | - | 21 | 2082 |