Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168319.1 | NP_001161791.1 | 4 | 753 | |
NM_001955.4 | NP_001946.3 | 4 | 756 | ||
XM_011514330.2 | XP_011512632.1 | 5 | 512 | ||
XM_011514331.2 | XP_011512633.1 | 6 | 673 | ||
XM_011514332.2 | XP_011512634.1 | 5 | 510 | ||
XM_017010331.1 | XP_016865820.1 | 5 | 696 | ||
GenBank mRNA | AK226096.1 | - | 4 | 671 | |
AK291838.1 | - | 4 | 693 | ||
AK313914.1 | - | 4 | 688 | ||
BC009720.1 | - | 4 | 625 | 3900704 | |
BX439515.2 | - | 4 | 716 | ||
CR541903.1 | - | 4 | 422 | ||
S56805.1 | - | 4 | 758 | ||
Y00749.1 | - | 4 | 674 |