Gene Transcripts
- Gene Symbol
- MYL6
- Entrez Gene ID
- 4637
- Gene Name
- myosin light chain 6
- Gene Aliases
- ESMLC, LC17, LC17-GI, LC17-NM, LC17A, LC17B, MLC-3, MLC1SM, MLC3NM, MLC3SM
- Chromosome Location
- Chr. 12: 56158261 - 56161582 on Build GRCh38
- UniGene ID
- Hs.632717
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021019.4 |
NP_066299.2
|
3, 4 |
325 |
|
NM_079423.3 |
NP_524147.2
|
3, 4 |
325 |
|
GenBank mRNA |
AB046613.1 |
- |
3, 4 |
224 |
|
AK301198.1 |
- |
2, 3 |
503 |
|
BC006781.2 |
- |
1, 2 |
155 |
|
BC017455.1 |
- |
3, 4 |
212 |
3538373 |
BC071661.1 |
- |
3, 4 |
208 |
5561575 |
BC093066.1 |
- |
3, 4 |
221 |
30722094 |
BG283793.1 |
- |
3, 4 |
325 |
|
CN401857.1 |
- |
3, 4 |
209 |
|
CR456839.1 |
- |
1, 2 |
79 |
|
CR457014.1 |
- |
2, 3 |
184 |
|
M22918.1 |
- |
3, 4 |
215 |
|
M22920.1 |
- |
2, 3 |
193 |
|
M31212.1 |
- |
3, 4 |
224 |
|
U02629.1 |
- |
3, 4 |
222 |
|
View Full Product Details