Gene Transcripts

Gene Symbol
RNF44
Entrez Gene ID
22838
Gene Name
ring finger protein 44
Gene Aliases
Chromosome Location
Chr. 5: 176526699 - 176543456 on Build GRCh38
UniGene ID
Hs.434888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014901.4 NP_055716.1 3 805
XM_005265840.2 XP_005265897.1 3 517
XM_005265841.4 XP_005265898.1 3 762
XM_005265842.4 XP_005265899.1 3 747
XM_005265843.2 XP_005265900.1 3 453
XM_005265844.3 XP_005265901.1 3 558
XM_005265845.4 XP_005265902.1 3 696
XM_006714831.2 XP_006714894.1 3 517
XM_006714832.2 XP_006714895.1 3 517
XM_011534466.2 XP_011532768.1 3 520
XM_011534467.2 XP_011532769.1 3 751
GenBank mRNA AB029023.1 - 3 690
AK302385.1 - 3 558
BC039833.1 - 3 761 5535911
BC063297.1 - 3 806 30349418

View Full Product Details