Gene Transcripts
- Gene Symbol
- PTP4A2
- Entrez Gene ID
- 8073
- Gene Name
- protein tyrosine phosphatase type IVA, member 2
- Gene Aliases
- HH13, HH7-2, HU-PP-1, OV-1, PRL-2, PRL2, PTP4A, PTPCAAX2, ptp-IV1a, ptp-IV1b
- Chromosome Location
- Chr. 1: 31906421 - 31942635 on Build GRCh38
- UniGene ID
- Hs.470477
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195100.1 |
NP_001182029.1
|
1, 2 |
687 |
|
NM_080391.3 |
NP_536316.1
|
2, 3 |
1088 |
|
XM_005271231.3 |
XP_005271288.1
|
2, 3 |
790 |
|
GenBank mRNA |
AF208850.1 |
- |
1, 2 |
435 |
|
AK292703.1 |
- |
1, 2 |
196 |
|
BC018662.1 |
- |
2, 3 |
1059 |
|
BC070182.1 |
- |
1, 2 |
422 |
4686913 |
L39000.1 |
- |
1, 2 |
57 |
|
U14603.1 |
- |
1, 2 |
517 |
|
U48297.1 |
- |
1, 2 |
159 |
|
View Full Product Details