Gene Transcripts

Gene Symbol
TUSC2
Entrez Gene ID
11334
Gene Name
tumor suppressor candidate 2
Gene Aliases
C3orf11, FUS1, PAP, PDAP2
Chromosome Location
Chr. 3: 50324909 - 50328238 on Build GRCh38
UniGene ID
Hs.517981
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007275.2 NP_009206.1 2 380
GenBank mRNA AF055479.1 - 2 388
AK311998.1 - 2 370
BC023976.2 - 2 332 3504783
CA392689.1 - 2 380

View Full Product Details