Gene Transcripts

Gene Symbol
C15orf40
Entrez Gene ID
123207
Gene Name
chromosome 15 open reading frame 40
Gene Aliases
Chromosome Location
Chr. 15: 82988297 - 83012298 on Build GRCh38
UniGene ID
Hs.352541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160113.1 NP_001153585.1 3 381
NM_001160114.1 NP_001153586.1 3 381
NM_001160115.1 NP_001153587.1 3 381
NM_001160116.1 NP_001153588.1 3 381
NM_144597.2 NP_653198.2 3 381
NR_027649.1 3 392
NR_027650.1 3 392
XM_006720385.2 XP_006720448.1 3 1038
XM_011521212.2 XP_011519514.1 3 1038
XM_011521213.2 XP_011519515.1 3 1038
XM_011521214.2 XP_011519516.1 3 1038
GenBank mRNA AJ705316.1 - 3 295
AK125586.1 - 2 1467
AK311484.1 - 3 343
AK312160.1 - 3 355
BC019820.1 - 3 346 4449390
BE906569.1 - 3 325
BQ185982.1 - 3 332
BY800475.1 - 3 314
CN347153.1 - 3 300
DC428269.1 - 3 379
KU178855.1 - 3 266

View Full Product Details