Gene Transcripts

Gene Symbol
C7orf31
Entrez Gene ID
136895
Gene Name
chromosome 7 open reading frame 31
Gene Aliases
Chromosome Location
Chr. 7: 25134697 - 25180748 on Build GRCh38
UniGene ID
Hs.122055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138811.3 NP_620166.3 3 840
XM_005249619.3 XP_005249676.1 3 689
XM_005249620.3 XP_005249677.1 4 1220
GenBank mRNA AK097248.1 - 4 561
BC043269.2 - 3 943 5297014
BC060879.1 - 3 753 5298874
BI830207.1 - 3 377

View Full Product Details