Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133264.4 | NP_573571.1 | 7 | 1485 | |
XM_005257083.2 | XP_005257140.1 | 7 | 1487 | ||
XM_005257084.2 | XP_005257141.1 | 7 | 1445 | ||
XM_011524412.1 | XP_011522714.1 | 6 | 1386 | ||
XM_011524413.1 | XP_011522715.1 | 6 | 1367 | ||
GenBank mRNA | AB043786.1 | - | 6 | 1245 | |
AF370396.1 | - | 7 | 1410 | ||
AJ431177.1 | - | 6 | 1245 | ||
AK026913.1 | - | 5 | 933 | ||
AK055057.1 | - | 5 | 702 | ||
AK289578.1 | - | 7 | 1445 | ||
AK289779.1 | - | 7 | 1504 | ||
AK290301.1 | - | 7 | 1481 | ||
AK315836.1 | - | 7 | 1465 | ||
AL834239.1 | - | 6 | 1354 | ||
BC025965.1 | - | 7 | 1441 | ||
BC065551.1 | - | 7 | 1422 | 6044222 |