Gene Transcripts
- Gene Symbol
- ALS2CR11
- Entrez Gene ID
- 151254
- Gene Name
- amyotrophic lateral sclerosis 2 chromosome region candidate 11
- Gene Aliases
- Chromosome Location
- Chr. 2: 201487421 - 201619182 on Build GRCh38
- UniGene ID
- Hs.335788
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168217.1 |
NP_001161689.1
|
12, 13 |
1258 |
|
NM_001168221.1 |
NP_001161693.1
|
12, 13 |
1258 |
|
NM_152525.5 |
NP_689738.3
|
12, 13 |
1258 |
|
XM_006712331.3 |
XP_006712394.1
|
12, 13 |
1261 |
|
XM_006712332.3 |
XP_006712395.1
|
12, 13 |
1261 |
|
XM_006712333.3 |
XP_006712396.1
|
12, 13 |
1261 |
|
XM_006712334.3 |
XP_006712397.1
|
12, 13 |
1261 |
|
XM_006712335.3 |
XP_006712398.1
|
12, 13 |
1261 |
|
GenBank mRNA |
AL833429.1 |
- |
12, 13 |
1255 |
|
BC030659.2 |
- |
12, 13 |
1251 |
4838801 |
BX648732.1 |
- |
12, 13 |
1251 |
|
View Full Product Details