Gene Transcripts

Gene Symbol
AAMDC
Entrez Gene ID
28971
Gene Name
adipogenesis associated Mth938 domain containing
Gene Aliases
C11orf67, CK067, PTD015
Chromosome Location
Chr. 11: 77821114 - 77901281 on Build GRCh38
UniGene ID
Hs.745040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316960.1 NP_001303889.1 5 578
NM_001316961.1 NP_001303890.1 5 498
NM_001316962.1 NP_001303891.1 5 502
NM_024684.3 NP_078960.1 4 460
XM_011544968.2 XP_011543270.1 4 460
XM_011544969.2 XP_011543271.1 4 453
GenBank mRNA AK024688.1 - 4 419
BC002752.2 - 4 412 3628061
BI791786.1 - 5 511
BM930331.1 - 5 447
CR457332.1 - 3 314

View Full Product Details