Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000597.2 | NP_000588.2 | 4 | 1222 | |
NM_001313990.1 | NP_001300919.1 | 4 | 780 | ||
NM_001313992.1 | NP_001300921.1 | 4 | 1099 | ||
NM_001313993.1 | NP_001300922.1 | 4 | 838 | ||
GenBank mRNA | A09809.1 | - | 4 | 1221 | |
AB209509.1 | - | 2 | 4208 | ||
BC004312.1 | - | 4 | 1196 | 3627826 | |
BC009902.1 | - | 4 | 1198 | 3509749 | |
BC012769.1 | - | 4 | 1196 | 3689990 | |
BC071967.1 | - | 4 | 1209 | 6650239 | |
X16302.1 | - | 4 | 1219 |