Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_201628.2 | NP_963922.2 | 14 | 2424 | |
XM_005245795.4 | XP_005245852.1 | 16 | 2679 | ||
XM_011541074.2 | XP_011539376.1 | 16 | 2686 | ||
XM_011541075.2 | XP_011539377.1 | 14 | 2342 | ||
XM_011541076.2 | XP_011539378.1 | 14 | 2487 | ||
XM_011541077.2 | XP_011539379.1 | 14 | 3466 | ||
XM_011541081.2 | XP_011539383.1 | 8 | 1166 | ||
GenBank mRNA | AK293806.1 | - | 9 | 1230 | |
BC101637.1 | - | 4 | 505 | ||
BC113621.1 | - | 4 | 505 | ||
BC143641.1 | - | 4 | 451 | ||
DA317442.1 | - | 4 | 442 | ||
DN831635.1 | - | 5 | 706 |