Gene Transcripts

Gene Symbol
MAGEA10
Entrez Gene ID
4109
Gene Name
MAGE family member A10
Gene Aliases
CT1.10, MAGE10
Chromosome Location
Chr. X: 152133310 - 152138578 on Build GRCh38
UniGene ID
Hs.18048
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011543.2 NP_001011543.2 5 481
NM_001251828.1 NP_001238757.1 5 409
NM_021048.4 NP_066386.2 4 390
GenBank mRNA AK314161.1 - 4 390
BC004105.2 - 4 365 3926146
CB158974.1 - 5 449
CB959556.1 - 5 492

View Full Product Details