Gene Transcripts

Gene Symbol
SIDT1
Entrez Gene ID
54847
Gene Name
SID1 transmembrane family member 1
Gene Aliases
SID-1, SID1
Chromosome Location
Chr. 3: 113531774 - 113629579 on Build GRCh38
UniGene ID
Hs.591291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308350.1 NP_001295279.1 11 1757
NM_001322294.1 NP_001309223.1 11 1757
NM_001322295.1 NP_001309224.1 10 1673
NM_001322296.1 NP_001309225.1 9 1571
NM_001322297.1 NP_001309226.1 10 1635
NM_001322298.1 NP_001309227.1 10 1734
NM_001322299.1 NP_001309228.1 11 1776
NM_001322300.1 NP_001309229.1 9 1571
NM_017699.2 NP_060169.2 11 1757
NR_136280.1 12 1837
NR_136281.1 10 1734
NR_136282.1 11 1757
NR_136283.1 11 1757
NR_136284.1 8 1527
NR_136285.1 8 1527
XM_011512939.2 XP_011511241.1 11 1717
XM_011512941.2 XP_011511243.1 11 1717
XM_017006664.1 XP_016862153.1 9 1648
XM_017006665.1 XP_016862154.1 9 2148
XM_017006666.1 XP_016862155.1 8 625
XM_017006667.1 XP_016862156.1 6 492
GenBank mRNA BC117222.1 - 11 1145
BC143776.1 - 11 1145
BC143778.1 - 11 1164

View Full Product Details