Gene Transcripts
- Gene Symbol
- CBWD1
- Entrez Gene ID
- 55871
- Gene Name
- COBW domain containing 1
- Gene Aliases
- COBP
- Chromosome Location
- Chr. 9: 121038 - 179083 on Build GRCh38
- UniGene ID
- Hs.531330
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145355.1 |
NP_001138827.1
|
8, 9 |
781 |
|
NM_001145356.1 |
NP_001138828.1
|
7, 8 |
683 |
|
NM_018491.3 |
NP_060961.3
|
7, 8 |
683 |
|
XM_005251513.4 |
XP_005251570.1
|
6, 7 |
631 |
|
XM_005251517.4 |
XP_005251574.1
|
7, 8 |
683 |
|
XM_011517961.2 |
XP_011516263.1
|
8, 9 |
775 |
|
XM_011517962.2 |
XP_011516264.1
|
7, 8 |
683 |
|
XM_011517963.1 |
XP_011516265.1
|
4, 5 |
221 |
|
XM_017014925.1 |
XP_016870414.1
|
7, 8 |
683 |
|
GenBank mRNA |
AF212253.1 |
- |
7, 8 |
651 |
|
BC005996.1 |
- |
7, 8 |
683 |
4069589 |
BC013432.1 |
- |
7, 8 |
654 |
4133722 |
View Full Product Details