Gene Transcripts

Gene Symbol
TTYH2
Entrez Gene ID
94015
Gene Name
tweety family member 2
Gene Aliases
C17orf29
Chromosome Location
Chr. 17: 74213557 - 74262020 on Build GRCh38
UniGene ID
Hs.27935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032646.5 NP_116035.5 14 1682
NM_052869.1 NP_443101.1 8 1492
XM_005257824.3 XP_005257881.1 14 1610
GenBank mRNA AF319952.1 - 14 1661
AK058197.1 - 3 382
AK091085.1 - 12 1363
AK126955.1 - 8 1900
AK296511.1 - 14 1605
AL831858.1 - 4 428
BC005168.1 - 14 1657
BC107492.1 - 14 1678 40033786
BC107602.1 - 14 1678 40033791
BX647912.1 - 8 1492

View Full Product Details