Gene Transcripts

Gene Symbol
KRTAP10-9
Entrez Gene ID
386676
Gene Name
keratin associated protein 10-9
Gene Aliases
KAP10.9, KAP18.9, KRTAP18-9
Chromosome Location
Chr. 21: 44627123 - 44628378 on Build GRCh38
UniGene ID
Hs.474000
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198690.2 NP_941963.2 1 526
GenBank mRNA AB076356.1 - 1 526
BC131613.1 - 1 502 40115414

View Full Product Details