Gene Transcripts

Gene Symbol
WTH3DI
Entrez Gene ID
150786
Gene Name
RAB6C-like
Gene Aliases
Chromosome Location
Chr. 2: 131360492 - 131364158 on Build GRCh38
UniGene ID
Hs.645670
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077637.1 NP_001071105.1 1 1277

View Full Product Details