Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001101417.3 | NP_001094887.1 | 9 | 1162 | |
NM_001101426.3 | NP_001094896.1 | 10 | 1312 | ||
XM_006715770.3 | XP_006715833.1 | 10 | 1482 | ||
XM_011515500.2 | XP_011513802.1 | 9 | 1352 | ||
XM_011515508.2 | XP_011513810.1 | 10 | 1814 | ||
XM_011515509.2 | XP_011513811.1 | 10 | 2663 | ||
XM_017012577.1 | XP_016868066.1 | 9 | 1574 | ||
XM_017012578.1 | XP_016868067.1 | 8 | 1989 | ||
GenBank mRNA | BE092109.1 | - | 3 | 253 |