Gene Transcripts

Gene Symbol
WAPL
Entrez Gene ID
23063
Gene Name
WAPL cohesin release factor
Gene Aliases
FOE, KIAA0261, WAPAL
Chromosome Location
Chr. 10: 86435256 - 86521816 on Build GRCh38
UniGene ID
Hs.203099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318328.1 NP_001305257.1 2 497
NM_015045.3 NP_055860.1 2 497
XM_011539548.1 XP_011537850.1 2 529
XM_017015979.1 XP_016871468.1 2 529
XM_017015980.1 XP_016871469.1 2 490
GenBank mRNA AB065003.1 - 2 500
AB426821.1 - 2 452
AB426822.1 - 4 402
AB426823.1 - 3 395
AB426824.1 - 3 319
AB426825.1 - 3 293
AB426826.1 - 3 282
AB426827.1 - 3 262
AB426828.1 - 3 214
AB426829.1 - 2 210
AB426830.1 - 2 204
AB426831.1 - 3 193
AB426832.1 - 2 191
AB426833.1 - 3 187
AB426834.1 - 3 175
AB426835.1 - 3 145
AB426836.1 - 2 118
AF479418.1 - 3 193
AK290138.1 - 2 458
BC140858.1 - 2 221 9021726
BC150269.1 - 3 348 9007232
D87450.1 - 3 348

View Full Product Details