Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130447.1 | NP_001123919.1 | 3 | 256 | |
NM_001323629.1 | NP_001310558.1 | 5 | 438 | ||
NM_001323631.1 | NP_001310560.1 | 5 | 438 | ||
NM_001323632.1 | NP_001310561.1 | 3 | 279 | ||
NM_018097.2 | NP_060567.1 | 4 | 349 | ||
XM_017022396.1 | XP_016877885.1 | 4 | 353 | ||
XM_017022397.1 | XP_016877886.1 | 4 | 320 | ||
GenBank mRNA | AK001322.1 | - | 4 | 307 | |
AK022939.1 | - | 4 | 320 | ||
BC010903.1 | - | 4 | 328 | 4285311 | |
DA923763.1 | - | 3 | 213 | ||
DB450370.1 | - | 4 | 349 |