Gene Transcripts
- Gene Symbol
- VWA5B1
- Entrez Gene ID
- 127731
- Gene Name
- von Willebrand factor A domain containing 5B1
- Gene Aliases
- Chromosome Location
- Chr. 1: 20290907 - 20360442 on Build GRCh38
- UniGene ID
- Hs.205178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039500.2 |
NP_001034589.2
|
7, 8 |
1169 |
|
XM_006710360.3 |
XP_006710423.2
|
4, 5 |
444 |
|
XM_011540683.2 |
XP_011538985.1
|
7, 8 |
1131 |
|
XM_011540684.2 |
XP_011538986.1
|
7, 8 |
1256 |
|
XM_011540685.2 |
XP_011538987.1
|
7, 8 |
1268 |
|
XM_011540688.2 |
XP_011538990.1
|
7, 8 |
1131 |
|
XM_011540691.2 |
XP_011538993.1
|
7, 8 |
1131 |
|
XM_011540692.2 |
XP_011538994.1
|
7, 8 |
1125 |
|
XM_011540693.1 |
XP_011538995.1
|
7, 8 |
1131 |
|
XM_011540695.1 |
XP_011538997.1
|
7, 8 |
1131 |
|
XM_011540698.1 |
XP_011539000.1
|
7, 8 |
1131 |
|
GenBank mRNA |
AK125833.1 |
- |
7, 8 |
1317 |
|
AK295797.1 |
- |
7, 8 |
1153 |
|
View Full Product Details