Gene Transcripts

Gene Symbol
PSMD13
Entrez Gene ID
5719
Gene Name
proteasome 26S subunit, non-ATPase 13
Gene Aliases
HSPC027, Rpn9, S11, p40.5
Chromosome Location
Chr. 11: 236675 - 252984 on Build GRCh38
UniGene ID
Hs.134688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002817.3 NP_002808.3 5 526
NM_175932.2 NP_787128.2 3 532
XM_011520235.2 XP_011518537.1 5 659
GenBank mRNA AB009398.1 - 5 353
AF083245.1 - 5 338
AF086708.1 - 5 284
AF107837.1 - 5 317
AK094775.1 - 3 363
AK222895.1 - 5 357
AK295944.1 - 4 323
AK307447.1 - 4 251
AK314733.1 - 5 788
BC001100.2 - 5 339 3506530
BC001747.1 - 5 307 3535807
BM456586.1 - 5 357
BT007307.1 - 5 284
BX647314.1 - 4 261

View Full Product Details