Gene Transcripts

Gene Symbol
NIF3L1
Entrez Gene ID
60491
Gene Name
NGG1 interacting factor 3 like 1
Gene Aliases
ALS2CR1, CALS-7, MDS015
Chromosome Location
Chr. 2: 200889327 - 200903932 on Build GRCh38
UniGene ID
Hs.145284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136039.2 NP_001129511.1 5 913
NM_001142355.1 NP_001135827.1 5 862
NM_021824.3 NP_068596.2 5 1065
XM_005246742.3 XP_005246799.1 5 1128
XM_011511582.1 XP_011509884.1 5 1049
XM_011511583.1 XP_011509885.1 5 1793
XM_011511584.1 XP_011509886.1 5 1611
XM_011511585.1 XP_011509887.1 5 1560
XM_017004630.1 XP_016860119.1 5 1310
GenBank mRNA AB038949.1 - 5 1026
AF060513.1 - 5 822
AF182416.1 - 5 987
AF283538.1 - 5 868
AK023378.1 - 5 1011
AK225905.1 - 5 2388
AK314709.1 - 5 867
AL834430.1 - 4 688
BC007654.2 - 4 849 3346099

View Full Product Details