Gene Transcripts

Gene Symbol
TRAPPC2
Entrez Gene ID
6399
Gene Name
trafficking protein particle complex 2
Gene Aliases
MIP2A, SEDL, SEDT, TRAPPC2P1, TRS20, ZNF547L, hYP38334
Chromosome Location
Chr. X: 13712242 - 13734635 on Build GRCh38
UniGene ID
Hs.592238
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011658.3 NP_001011658.1 3 320
NM_001128835.2 NP_001122307.2 3 314
NM_014563.5 NP_055378.1 2 178
XM_011545565.1 XP_011543867.1 4 388
XM_011545566.2 XP_011543868.1 3 244
GenBank mRNA AK098253.1 - 1 2327
AK310542.1 - 3 224
AK313736.1 - 3 260
BC016915.1 - 2 141 4432196
BC052618.1 - 2 141 6581382
BI821518.1 - 2 152
CB962763.1 - 2 163
CD698535.1 - 3 274
DA344866.1 - 2 178
DA350753.1 - 3 308
DA542477.1 - 3 254

Gene Symbol
TRAPPC2B
Entrez Gene ID
10597
Gene Name
trafficking protein particle complex 2B
Gene Aliases
MIP-2A, SEDLP, SEDLP1, TRAPPC2P1
Chromosome Location
Chr. 19: 57363511 - 57365353 on Build GRCh38
UniGene ID
Hs.446620
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002166.1 2 260
GenBank mRNA AF058918.1 - 2 260
BU556832.1 - 2 254
CR456859.1 - 1 48

View Full Product Details