Gene Transcripts

Gene Symbol
KY
Entrez Gene ID
339855
Gene Name
kyphoscoliosis peptidase
Gene Aliases
Chromosome Location
Chr. 3: 134599923 - 134656053 on Build GRCh38
UniGene ID
Hs.146730
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178554.4 NP_848649.3 2, 3 254
XM_017006287.1 XP_016861776.1 2, 3 5285
XM_017006288.1 XP_016861777.1 2, 3 5231
GenBank mRNA AK090526.1 - 2, 3 254
AK294500.1 - 2, 3 255
BC136349.1 - 2, 3 215
DA238247.1 - 2, 3 254

View Full Product Details