Gene Transcripts

Gene Symbol
SMIM4
Entrez Gene ID
440957
Gene Name
small integral membrane protein 4
Gene Aliases
C3orf78
Chromosome Location
Chr. 3: 52535986 - 52540570 on Build GRCh38
UniGene ID
Hs.660577
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001124767.1 NP_001118239.1 2 356
XM_011533729.2 XP_011532031.1 2 472
GenBank mRNA BC017996.1 - 2 356
BG031013.1 - 2 346

View Full Product Details