Gene Transcripts

Gene Symbol
TP73-AS1
Entrez Gene ID
57212
Gene Name
TP73 antisense RNA 1
Gene Aliases
KIAA0495, PDAM
Chromosome Location
Chr. 1: 3735984 - 3747373 on Build GRCh38
UniGene ID
Hs.49658
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033710.1 4 2220
NR_033711.1 4 2220
GenBank mRNA AB007964.1 - 4 2170
AL133648.1 - 4 1473

View Full Product Details