Gene Transcripts
- Gene Symbol
- COLGALT2
- Entrez Gene ID
- 23127
- Gene Name
- collagen beta(1-O)galactosyltransferase 2
- Gene Aliases
- C1orf17, GLT25D2
- Chromosome Location
- Chr. 1: 183929662 - 184051612 on Build GRCh38
- UniGene ID
- Hs.387995
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303420.1 |
NP_001290349.1
|
10, 11 |
1810 |
|
NM_001303421.1 |
NP_001290350.1
|
10, 11 |
1324 |
|
NM_015101.3 |
NP_055916.1
|
10, 11 |
1810 |
|
XM_011509332.2 |
XP_011507634.1
|
11, 12 |
1616 |
|
XM_011509334.2 |
XP_011507636.1
|
11, 12 |
1423 |
|
XM_011509335.2 |
XP_011507637.1
|
11, 12 |
1474 |
|
XM_011509336.2 |
XP_011507638.1
|
11, 12 |
1251 |
|
XM_011509337.2 |
XP_011507639.1
|
7, 8 |
1550 |
|
XM_011509338.2 |
XP_011507640.1
|
8, 9 |
1302 |
|
XM_017000750.1 |
XP_016856239.1
|
10, 11 |
1375 |
|
XM_017000751.1 |
XP_016856240.1
|
5, 6 |
1234 |
|
GenBank mRNA |
AB011156.1 |
- |
10, 11 |
1735 |
|
AF288389.1 |
- |
10, 11 |
1769 |
|
AK095090.1 |
- |
5, 6 |
1045 |
|
AK095192.1 |
- |
10, 11 |
1298 |
|
AK127287.1 |
- |
2, 3 |
1315 |
|
AK293973.1 |
- |
10, 11 |
1517 |
|
BC035672.1 |
- |
10, 11 |
1792 |
5740842 |
View Full Product Details