Gene Transcripts

Gene Symbol
Adam11
Entrez Gene ID
11488
Gene Name
a disintegrin and metallopeptidase domain 11
Gene Aliases
AW060611, Mdc
Chromosome Location
Chr. 11: 102761439 - 102780262 on Build GRCm38
UniGene ID
Mm.89854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110778.1 NP_001104248.1 14, 15 1411
NM_009613.2 NP_033743.2 14, 15 1411
GenBank mRNA AB009676.1 - 14, 15 1244
AK157926.1 - 14, 15 1416
BC054536.1 - 14, 15 1271 6400084

View Full Product Details