Gene Transcripts

Gene Symbol
Speer4a
Entrez Gene ID
75657
Gene Name
spermatogenesis associated glutamate (E)-rich protein 4a
Gene Aliases
1700027N01Rik, SPEER-4A
Chromosome Location
Chr. 5: 26017738 - 26042295 on Build GRCm38
UniGene ID
Mm.374273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029376.2 NP_083652.2 4 597
GenBank mRNA AK006439.1 - 4 601

View Full Product Details