Gene Transcripts

Gene Symbol
Klk1b26
Entrez Gene ID
16618
Gene Name
kallikrein 1-related petidase b26
Gene Aliases
Egfbp2, Klk26, PRECE-2, mGK-26, mGK26
Chromosome Location
Chr. 7: 44007953 - 44016969 on Build GRCm38
UniGene ID
Mm.439663
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010644.2 NP_034774.1 4 609
GenBank mRNA AK075752.1 - 2 165
AK075766.1 - 2 165
BC024688.1 - 4 583 4167696
BC138704.1 - 4 591 8861726
K01831.1 - 4 609
X63327.1 - 4 588

Gene Symbol
Egfbp2
Entrez Gene ID
13647
Gene Name
epidermal growth factor binding protein type B
Gene Aliases
Egfbp-2, Klk1b26, mGk-13
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.439663
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010115.6 NP_034245.3 4 591
GenBank mRNA AB016032.1 - 4 567
X58628.1 - 4 591

View Full Product Details