Gene Transcripts

Gene Symbol
Rex2
Entrez Gene ID
100043034
Gene Name
reduced expression 2
Gene Aliases
Gm13138, OTTMUSG00000010655
Chromosome Location
Chr. 4: 147021850 - 147060799 on Build GRCm38
UniGene ID
Mm.422606
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177767.1 NP_001171238.1 5 963
GenBank mRNA BC067397.1 - 3 718

View Full Product Details