Gene Transcripts

Gene Symbol
Rc3h2
Entrez Gene ID
319817
Gene Name
ring finger and CCCH-type zinc finger domains 2
Gene Aliases
2900024N03Rik, 9430019J22Rik, D930043C02Rik, Mnab, Rnf164
Chromosome Location
Chr. 2: 37368861 - 37423380 on Build GRCm38
UniGene ID
Mm.288785
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100591.1 NP_001094061.1 22 3743
NM_001290642.1 NP_001277571.1 22 3724
GenBank mRNA AK053071.1 - 22 3724
BC117951.1 - 22 3736 40098089
BC117952.1 - 22 3736 40098090

View Full Product Details