Gene Transcripts

Gene Symbol
Klf14
Entrez Gene ID
619665
Gene Name
Kruppel-like factor 14
Gene Aliases
5330411L03Rik, AL022736, BTEB5
Chromosome Location
Chr. 6: 30956021 - 30958990 on Build GRCm38
UniGene ID
Mm.204671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135093.1 NP_001128565.1 1 998
GenBank mRNA AK030435.1 - 2 292
DQ534758.1 - 1 990

View Full Product Details