Gene Transcripts

Gene Symbol
Ier2
Entrez Gene ID
15936
Gene Name
immediate early response 2
Gene Aliases
AI317238, Ch1, Pip92
Chromosome Location
Chr. 8: 84661331 - 84662852 on Build GRCm38
UniGene ID
Mm.399
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010499.4 NP_034629.3 1 1491
GenBank mRNA AK032692.1 - 1489
AK144601.1 - 1 1488
AK150860.1 - 1 1486
AK152747.1 - 1 1486
AK170433.1 - 1488
AK172332.1 - 1 1431
BC002067.1 - 1 1476 3590296
M31042.1 - 1 1488
M59821.1 - 1 1491

View Full Product Details