Gene Transcripts

Gene Symbol
Vps13b
Entrez Gene ID
666173
Gene Name
vacuolar protein sorting 13B (yeast)
Gene Aliases
1810042B05Rik, 2310042E16Rik, 4732488H20, C330002D13Rik, C87206, Coh1, D230005K13, mKIAA0532, mKIAA5032
Chromosome Location
Chr. 15: 35369882 - 35931229 on Build GRCm38
UniGene ID
Mm.309250
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177151.3 NP_796125.2 24 3893
GenBank mRNA AK009750.1 - 4 625
BC089558.1 - 4 642
BC158052.1 - 24 3873

View Full Product Details