Gene Transcripts

Gene Symbol
Sipa1l3
Entrez Gene ID
74206
Gene Name
signal-induced proliferation-associated 1 like 3
Gene Aliases
2610511M17Rik
Chromosome Location
Chr. 7: 29320372 - 29518943 on Build GRCm38
UniGene ID
Mm.447042
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081028.1 NP_001074497.1 12, 13 3995
GenBank mRNA AK154121.1 - 12, 13 3991
AK163313.1 - 9, 10 2377
BC095969.1 - 2, 3 528

View Full Product Details