Gene Transcripts

Gene Symbol
Sipa1l3
Entrez Gene ID
74206
Gene Name
signal-induced proliferation-associated 1 like 3
Gene Aliases
2610511M17Rik
Chromosome Location
Chr. 7: 29320372 - 29518943 on Build GRCm38
UniGene ID
Mm.447042
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081028.1 NP_001074497.1 5 2233
GenBank mRNA AK154121.1 - 5 2234
AK163313.1 - 2 614
AK171182.1 - 4 2157

View Full Product Details