Gene Transcripts

Gene Symbol
Zscan4c
Entrez Gene ID
245109
Gene Name
zinc finger and SCAN domain containing 4C
Gene Aliases
Gm397, XM_142517, Zscan4d
Chromosome Location
Chr. 7: 11005745 - 11010547 on Build GRCm38
UniGene ID
Mm.131747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013765.2 NP_001013787.1 4 2152
GenBank mRNA BC050218.1 - 4 2152

View Full Product Details