Gene Transcripts

Gene Symbol
Vmn2r49
Entrez Gene ID
625605
Gene Name
vomeronasal 2, receptor 49
Gene Aliases
Chromosome Location
Chr. 7: 9976245 - 9992139 on Build GRCm38
UniGene ID
Mm.484763
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105156.1 NP_001098626.1 6 2314

View Full Product Details