Gene Transcripts
- Gene Symbol
- Gpr27
- Entrez Gene ID
- 14761
- Gene Name
- G protein-coupled receptor 27
- Gene Aliases
- Chromosome Location
- Chr. 6: 99692679 - 99693818 on Build GRCm38
- UniGene ID
- Mm.35009
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008158.1 |
NP_032184.1
|
1 |
1038 |
|
View Full Product Details