Gene Transcripts

Gene Symbol
Rhox6
Entrez Gene ID
19202
Gene Name
reproductive homeobox 6
Gene Aliases
AV213605, AW548842, Psx1
Chromosome Location
Chr. X: 37827055 - 37829857 on Build GRCm38
UniGene ID
Mm.10105
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008955.1 NP_032981.1 2 453
GenBank mRNA AF017453.1 - 2 453
AK076186.1 - 2 462
BC099477.1 - 2 449 30790014
DQ058643.1 - 2 453

View Full Product Details