Gene Transcripts

Gene Symbol
Tbc1d4
Entrez Gene ID
210789
Gene Name
TBC1 domain family, member 4
Gene Aliases
5930406J04Rik, A930035N22, AV295684, As160
Chromosome Location
Chr. 14: 101442360 - 101609228 on Build GRCm38
UniGene ID
Mm.320639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081278.2 NP_001074747.2 11 2829
GenBank mRNA AK031120.1 - 11 2673
AK039337.1 - 13 3019
AK044719.1 - 11 2471
AK220347.1 - 10 1236
BC042747.1 - 7 831
BC064433.1 - 11 2453
BC080762.1 - 11 2447
BC117869.1 - 11 2302 40095526
BC117870.1 - 12 2329 40095531

View Full Product Details