Gene Transcripts

Gene Symbol
CAMTA1
Entrez Gene ID
23261
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
CANPMR
Chromosome Location
Chr. 1: 6785324 - 7769706 on Build GRCh38
UniGene ID
Hs.397705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015215.3 NP_056030.1 11, 12 3115
XM_011541083.2 XP_011539385.1 11, 12 2979
XM_011541084.2 XP_011539386.1 11, 12 2979
XM_011541085.2 XP_011539387.1 12, 13 3051
XM_011541086.2 XP_011539388.1 11, 12 2979
XM_011541087.2 XP_011539389.1 10, 11 2907
XM_011541088.2 XP_011539390.1 10, 11 2909
XM_011541089.2 XP_011539391.1 11, 12 2979
XM_011541090.2 XP_011539392.1 11, 12 2979
XM_017000773.1 XP_016856262.1 12, 13 3731
XM_017000774.1 XP_016856263.1 11, 12 2979
XM_017000775.1 XP_016856264.1 10, 11 2909
XM_017000776.1 XP_016856265.1 13, 14 3111
XM_017000777.1 XP_016856266.1 11, 12 2979
XM_017000778.1 XP_016856267.1 11, 12 2979
XM_017000779.1 XP_016856268.1 10, 11 2909
GenBank mRNA AB020640.3 - 11, 12 3115
AK302088.1 - 2, 3 241
BC151835.1 - 11, 12 3115 8860115

View Full Product Details