Gene Transcripts

Gene Symbol
Scand1
Entrez Gene ID
19018
Gene Name
SCAN domain-containing 1
Gene Aliases
2310003H23Rik, Leap1, PGC-2, Ppargc2
Chromosome Location
Chr. 2: 156311846 - 156312704 on Build GRCm38
UniGene ID
Mm.389346
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020255.3 NP_064651.2 1 132

View Full Product Details