Gene Transcripts

Gene Symbol
TSPYL4
Entrez Gene ID
23270
Gene Name
TSPY like 4
Gene Aliases
dJ486I3.2
Chromosome Location
Chr. 6: 116249964 - 116254098 on Build GRCh38
UniGene ID
Hs.284141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021648.4 NP_067680.3 1 859
GenBank mRNA AB018264.1 - 1 828
AK091559.1 - 3 612
AK302542.1 - 1 862
BC009116.1 - 1 233
BC040453.1 - 1 803

View Full Product Details