Gene Transcripts

Gene Symbol
Spatc1
Entrez Gene ID
315091
Gene Name
spermatogenesis and centriole associated 1
Gene Aliases
RGD1565316
Chromosome Location
Chr. 7: 117326279 - 117349957 on Build Rnor_6.0
UniGene ID
Rn.172256
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115026.1 NP_001108498.1 3 1007
GenBank mRNA BC083658.1 - 3 1091
BC103732.1 - 3 1090
BC158623.1 - 3 1002 7454231

View Full Product Details